1-(3-Chloro-2-methylphenyl)-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264706-5G 264706-25G 264706-1G 264706-500MG
77632-358EA 776 USD
77632-358 77654-556 77641-614 77645-564
1-(3-Chloro-2-methylphenyl)-2-methyl-1-propanone ≥95%
1-(3-Chloro-2-methylphenyl)-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: 196.67 g/mol
MW: C₁₁H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD19314973
CAS Number: 1267143-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR