1-(2,4-Dichlorophenyl)-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264647-5G 264647-1G 264647-25G 264647-500MG
77632-356EA 1327 USD
77632-356 77637-582 77658-324 77649-658
1-(2,4-Dichlorophenyl)-2-methyl-1-propanone ≥95%
1-(2,4-Dichlorophenyl)-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: C₁₀H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD14609629
CAS Number: 107680-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR