1-(5-Fluoro-2-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264590-5G 264590-25G 264590-1G 264590-500MG
77632-352EA 1011 USD
77632-352 77658-318 77641-610 77645-560
1-(5-Fluoro-2-methylphenyl)-1-propanone ≥95%
1-(5-Fluoro-2-methylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 166.19 g/mol
MW: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD11617672
CAS Number: 1250903-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR