1-(5-Chloro-2-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264585-5G 264585-1G 264585-25G 264585-500MG
77632-350EA 659 USD
77632-350 77637-576 77654-552 77645-558
1-(5-Chloro-2-methylphenyl)-1-propanone ≥95%
1-(5-Chloro-2-methylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 182.65 g/mol
MW: C₁₀H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD11617659
CAS Number: 1193779-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR