1-(5-Bromo-2-fluorophenyl)propan-1-one ≥97%

Supplier: AOBChem USA

Warning

264578-5G 264578-1G 264578-25G 264578-500MG
77634-544EA 285 USD
77634-544 77637-572 77658-314 77645-554
1-(5-Bromo-2-fluorophenyl)propan-1-one ≥97%
1-(5-Bromo-2-fluorophenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₉H₈BrFO
MW: 231.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226832
CAS Number: 864774-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR