2-Methyl-1-(3-(trifluoromethyl)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

264567-5G 264567-25G
77634-542EA 151 USD
77634-542 77654-550
2-Methyl-1-(3-(trifluoromethyl)phenyl)propan-1-one ≥95%
2-Methyl-1-(3-(trifluoromethyl)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₁₁H₁₁F₃O
MW: 216.2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00458543
CAS Number: 586369-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR