1-(2,5-Dimethylphenyl)-2-methyl-1-propanone ≥97%

Supplier: AOBChem USA

Warning

264551-5G 264551-25G 264551-1G 264551-500MG
77632-346EA 1159 USD
77632-346 77658-312 77641-608 77649-654
1-(2,5-Dimethylphenyl)-2-methyl-1-propanone ≥97%
1-(2,5-Dimethylphenyl)-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: 176.26 g/mol
MW: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD09891212
CAS Number: 5445-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR