1-(2-Fluoro-4-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264505-5G 264505-1G 264505-500MG
77628-304EA 1484.23 USD
77628-304 77637-570 77649-650
1-(2-Fluoro-4-methylphenyl)-1-propanone ≥95%
1-(2-Fluoro-4-methylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 166.19 g/mol
MW: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD18397081
CAS Number: 107076-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR