[3-Bromo-4-(propan-2-yloxy)phenyl]methanol ≥95%

Supplier: AOBChem USA

Warning

26431-5G 26431-1G 26431-25G 26431-500MG
77632-342EA 784 USD
77632-342 77637-568 77654-546 77649-648
[3-Bromo-4-(propan-2-yloxy)phenyl]methanol ≥95%
[3-Bromo-4-(propan-2-yloxy)phenyl]methanol

Precaución: For Research Use Only

Formula: 245.113 g/mol
MW: C₁₀H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD09754609
CAS Number: 454186-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR