(3,5-Dibromothiophen-2-yl)boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

26384-5G 26384-25G 26384-1G 26384-500MG
77632-340EA 692 USD
77632-340 77658-310 77641-604 77649-646
(3,5-Dibromothiophen-2-yl)boronic acid pinacol ester ≥95%
(3,5-Dibromothiophen-2-yl)boronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₀H₁₃BBr₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD26045276
CAS Number: 2121511-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR