3,4-Difluorobenzhydrazide ≥97%

Supplier: AOBChem USA

Warning

263710-5G 263710-25G
77634-540EA 86 USD
77634-540 77654-544
3,4-Difluorobenzhydrazide ≥97%
3,4-Difluorobenzhydrazide

Precaución: For Research Use Only

Formula: C₇H₆F₂N₂O
MW: 172.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672992
CAS Number: 229957-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR