3-Bromo-5-methylbenzyl bromide ≥95%

Supplier: AOBChem USA

Warning

26331-5G 26331-1G 26331-25G 26331-500MG
77634-536EA 418 USD
77634-536 77637-562 77658-304 77649-636
3-Bromo-5-methylbenzyl bromide ≥95%
3-Bromo-5-methylbenzyl bromide

Precaución: For Research Use Only

Formula: C₈H₈Br₂
MW: 263.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18204792
CAS Number: 51719-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR