1-(2-Ethyl-5-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

263065-5G 263065-25G 263065-1G 263065-500MG
77628-300EA 1703.62 USD
77628-300 77658-302 77641-598 77649-634
1-(2-Ethyl-5-fluorophenyl)ethanone ≥95%
1-(2-Ethyl-5-fluorophenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD28552602
CAS Number: 1785133-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR