2-Ethyl-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

263055-1G 263055-5G 263055-500MG
77634-534EA 823 USD
77634-534 77658-300 77649-632
2-Ethyl-5-fluorobenzoic acid ≥97%
2-Ethyl-5-fluorobenzoic acid

Precaución: For Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD18411932
CAS Number: 959992-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR