1-Ethyl-4-fluoro-2-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

263032-5G 263032-1G 263032-25G 263032-500MG
77628-298EA 1484.23 USD
77628-298 77637-560 77658-298 77645-548
1-Ethyl-4-fluoro-2-(methylthio)benzene ≥95%
1-Ethyl-4-fluoro-2-(methylthio)benzene

Precaución: For Research Use Only

Formula: 170.25 g/mol
MW: C₉H₁₁FS
Storage Temperature: Ambient
MDL Number: MFCD22489279
CAS Number: 1824369-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR