4'-Bromo-3-methoxy-1,1'-biphenyl ≥95%
Supplier: AOBChem USA
|
Warning
|
262098-5G
262098-25G
262098-1G
262098-500MG
77632-332EA
998
USD
77632-332
77658-296
77641-592
77649-624
4'-Bromo-3-methoxy-1,1'-biphenyl ≥95%
4'-Bromo-3-methoxy-1,1'-biphenyl
Precaución: For Research Use Only
Formula:
C₁₃H₁₁BrO Storage Temperature: Ambient |
MDL Number:
MFCD06201351 CAS Number: 74447-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...