Methyl β-oxo-2-(trifluoromethoxy)benzenepropanoate ≥95%

Supplier: AOBChem USA

Warning

261543-5G 261543-1G 261543-500MG
77628-290EA 1484.23 USD
77628-290 77641-588 77649-620
Methyl β-oxo-2-(trifluoromethoxy)benzenepropanoate ≥95%
Methyl β-oxo-2-(trifluoromethoxy)benzenepropanoate

Precaución: For Research Use Only

Formula: 262.18 g/mol
MW: C₁₁H₉F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD22665928
CAS Number: 1435442-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR