4-Bromo-1-ethyl-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

26148-5G 26148-25G 26148-1G 26148-500MG
77632-328EA 1268 USD
77632-328 77658-292 77641-586 77645-536
4-Bromo-1-ethyl-2-iodobenzene ≥97%
4-Bromo-1-ethyl-2-iodobenzene

Precaución: For Research Use Only

Formula: C₈H₈BrI
Storage Temperature: Ambient
MDL Number: MFCD22490701
CAS Number: 1033760-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR