2,5-Dibromo-4-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

261408-5G 261408-1G 261408-500MG 261408-25G
77632-324EA 679 USD
77632-324 77637-548 77652-548 77654-526
2,5-Dibromo-4-fluoropyridine ≥97%
2,5-Dibromo-4-fluoropyridine

Precaución: For Research Use Only

Formula: C₅H₂Br₂FN
MW: 254.88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100660
CAS Number: 1033203-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR