2-(Cyclohexyloxy)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

260818-5G 260818-25G 260818-1G 260818-500MG
77634-528EA 601 USD
77634-528 77654-522 77641-582 77645-534
2-(Cyclohexyloxy)benzonitrile ≥97%
2-(Cyclohexyloxy)benzonitrile

Precaución: For Research Use Only

Formula: 201.26 g/mol
MW: C₁₃H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD11155921
CAS Number: 1020920-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR