2-(Cyclohexyloxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

260816-5G 260816-1G 260816-25G 260816-500MG
77632-322EA 822 USD
77632-322 77637-546 77658-288 77649-616
2-(Cyclohexyloxy)benzoic acid ≥95%
2-(Cyclohexyloxy)benzoic acid

Precaución: For Research Use Only

Formula: 220.26 g/mol
MW: C₁₃H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD11156584
CAS Number: 55751-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR