1-Bromo-2-(cyclohexyloxy)benzene ≥95%

Supplier: AOBChem USA

Warning

260815-5G 260815-25G 260815-1G 260815-500MG
77634-526EA 334 USD
77634-526 77658-286 77641-580 77649-614
1-Bromo-2-(cyclohexyloxy)benzene ≥95%
1-Bromo-2-(cyclohexyloxy)benzene

Precaución: For Research Use Only

Formula: C₁₂H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD11167623
CAS Number: 105902-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR