1-(Cyclohexyloxy)-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

260813-5G 260813-1G 260813-25G 260813-500MG
77634-524EA 556 USD
77634-524 77637-544 77658-284 77645-532
1-(Cyclohexyloxy)-2-nitrobenzene ≥97%
1-(Cyclohexyloxy)-2-nitrobenzene

Precaución: For Research Use Only

Formula: C₁₂H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD13689263
CAS Number: 30718-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR