3-Chloro-2-(cyclopropylmethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

260402-5G 260402-25G 260402-1G 260402-500MG
77632-320EA 1140 USD
77632-320 77654-520 77641-578 77645-530
3-Chloro-2-(cyclopropylmethoxy)pyridine ≥97%
3-Chloro-2-(cyclopropylmethoxy)pyridine

Precaución: For Research Use Only

Formula: 183.64 g/mol
MW: C₉H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD28128666
CAS Number: 1355068-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR