2,2'-Dichloro[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

260188-5G 260188-25G 260188-1G 260188-500MG
77628-288EA 1484.23 USD
77628-288 77658-282 77641-576 77645-528
2,2'-Dichloro[1,1'-biphenyl]-4-carboxylic acid ≥97%
2,2'-Dichloro[1,1'-biphenyl]-4-carboxylic acid

Precaución: For Research Use Only

Formula: C₁₃H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD23549202
CAS Number: 627910-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR