5-Bromo-N-phenylpyridin-3-amine ≥97%

Supplier: AOBChem USA

Warning

26016-5G 26016-25G 26016-1G 26016-500MG
77632-318EA 841 USD
77632-318 77658-280 77641-574 77645-526
5-Bromo-N-phenylpyridin-3-amine ≥97%
5-Bromo-N-phenylpyridin-3-amine

Precaución: For Research Use Only

Formula: 249.107 g/mol
MW: C₁₁H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD14702686
CAS Number: 767342-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR