1-(2-Fluoro-5-methylpyridin-3-yl)ethan-1-ol ≥97%

Supplier: AOBChem USA

Warning

26008-5G 26008-1G 26008-500MG 26008-25G
77632-314EA 656 USD
77632-314 77637-540 77652-546 77658-276
1-(2-Fluoro-5-methylpyridin-3-yl)ethan-1-ol ≥97%
1-(2-Fluoro-5-methylpyridin-3-yl)ethan-1-ol

Precaución: For Research Use Only

Formula: C₈H₁₀FNO
MW: 155.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24848704
CAS Number: 1449008-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR