2,6-Dichloro-N-methoxy-N-methylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

25995-5G 25995-25G 25995-1G 25995-500MG
77634-522EA 263 USD
77634-522 77658-274 77641-572 77649-610
2,6-Dichloro-N-methoxy-N-methylnicotinamide ≥97%
2,6-Dichloro-N-methoxy-N-methylnicotinamide

Precaución: For Research Use Only

Formula: C₈H₈Cl₂N₂O₂
MW: 235.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847422
CAS Number: 873936-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR