α-Cyclopropyl-3-(trifluoromethoxy)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259533-5G 259533-1G 259533-500MG
77658-272EA 1898 USD
77658-272 77641-570 77649-608
α-Cyclopropyl-3-(trifluoromethoxy)benzenemethanol ≥95%
α-Cyclopropyl-3-(trifluoromethoxy)benzenemethanol

Precaución: For Research Use Only

Formula: 232.2 g/mol
MW: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD17248277
CAS Number: 1343341-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR