α-Cyclopropyl-3-fluoro-5-(trifluoromethyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

259531-5G 259531-1G 259531-500MG
77654-518EA 2245 USD
77654-518 77641-568 77645-524
α-Cyclopropyl-3-fluoro-5-(trifluoromethyl)benzenemethanol ≥97%
α-Cyclopropyl-3-fluoro-5-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 234.19 g/mol
MW: C₁₁H₁₀F₄O
Storage Temperature: Ambient
MDL Number: MFCD24185492
CAS Number: 1546689-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR