3-Chloro-α-cyclopropyl-4-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259509-5G 259509-500MG 259509-25G 259509-1G
77628-284EA 1484.23 USD
77628-284 77652-544 77654-516 77641-564
3-Chloro-α-cyclopropyl-4-methoxybenzenemethanol ≥95%
3-Chloro-α-cyclopropyl-4-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: 212.67 g/mol
MW: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD12786782
CAS Number: 1216073-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR