α-Cyclopropyl-4-fluoro-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

259493-5G 259493-1G 259493-500MG
77628-280EA 1484.23 USD
77628-280 77641-562 77649-606
α-Cyclopropyl-4-fluoro-1-naphthalenemethanol ≥95%
α-Cyclopropyl-4-fluoro-1-naphthalenemethanol

Precaución: For Research Use Only

Formula: C₁₄H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD19653939
CAS Number: 1343967-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR