1-Bromo-3-fluoro-4-iodo-2-(methoxymethoxy)benzene ≥95%
Supplier: AOBChem USA
|
Warning
|
25949-5G
25949-1G
25949-25G
25949-500MG
77628-282EA
1484.23
USD
77628-282
77637-538
77654-514
77645-520
1-Bromo-3-fluoro-4-iodo-2-(methoxymethoxy)benzene ≥95%
1-Bromo-3-fluoro-4-iodo-2-(methoxymethoxy)benzene
Precaución: For Research Use Only
Formula:
360.947 g/mol MW: C₈H₇BrFIO₂ Storage Temperature: Ambient |
MDL Number:
MFCD34755816 CAS Number: 2901109-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...