5-Chloro-α-cyclopropyl-2-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

259475-5G 259475-1G 259475-500MG
77654-512EA 1898 USD
77654-512 77641-560 77645-518
5-Chloro-α-cyclopropyl-2-fluorobenzenemethanol ≥97%
5-Chloro-α-cyclopropyl-2-fluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD34875832
CAS Number: 1602128-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR