4-Chloro-α-cyclopropyl-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259472-5G 259472-1G 259472-500MG
77628-278EA 1817.95 USD
77628-278 77637-536 77649-604
4-Chloro-α-cyclopropyl-2-methoxybenzenemethanol ≥95%
4-Chloro-α-cyclopropyl-2-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD29948575
CAS Number: 1603571-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR