3-Chloro-α-cyclopropyl-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259461-5G 259461-1G 259461-500MG
77628-276EA 1484.23 USD
77628-276 77637-534 77649-602
3-Chloro-α-cyclopropyl-2-methoxybenzenemethanol ≥95%
3-Chloro-α-cyclopropyl-2-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34875825
CAS Number: 2353136-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR