α-Cyclopropyl-3-fluoro-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259456-1G 259456-5G 259456-500MG
77637-532EA 549 USD
77637-532 77658-268 77645-514
α-Cyclopropyl-3-fluoro-2-methoxybenzenemethanol ≥95%
α-Cyclopropyl-3-fluoro-2-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: 196.22 g/mol
MW: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34875823
CAS Number: 2353830-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR