α-Cyclopropyl-3-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259446-5G 259446-1G 259446-25G 259446-500MG
77632-312EA 1206 USD
77632-312 77637-530 77658-266 77649-596
α-Cyclopropyl-3-(trifluoromethyl)benzenemethanol ≥95%
α-Cyclopropyl-3-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 216.2 g/mol
MW: C₁₁H₁₁F₃O
Storage Temperature: Ambient
MDL Number: MFCD16293523
CAS Number: 6552-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR