α-Cyclopropyl-2,3-difluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

259440-5G 259440-1G 259440-500MG
77628-272EA 1484.23 USD
77628-272 77645-512 77649-594
α-Cyclopropyl-2,3-difluorobenzenemethanol ≥97%
α-Cyclopropyl-2,3-difluorobenzenemethanol

Precaución: For Research Use Only

Formula: 184.18 g/mol
MW: C₁₀H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD19626916
CAS Number: 1341584-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR