2-Chloro-α-cyclopropyl-4-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259422-5G 259422-1G 259422-500MG
77628-270EA 1667.57 USD
77628-270 77641-552 77645-508
2-Chloro-α-cyclopropyl-4-fluorobenzenemethanol ≥95%
2-Chloro-α-cyclopropyl-4-fluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD20335997
CAS Number: 1202774-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR