α-Cyclopropyl-2-fluoro-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259408-5G 259408-1G 259408-500MG
77628-264EA 1599.59 USD
77628-264 77637-524 77649-588
α-Cyclopropyl-2-fluoro-4-methylbenzenemethanol ≥95%
α-Cyclopropyl-2-fluoro-4-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD34147376
CAS Number: 1593406-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR