α-Cyclopropyl-3-methoxy-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259398-5G 259398-1G 259398-500MG
77628-262EA 1707.74 USD
77628-262 77641-548 77649-586
α-Cyclopropyl-3-methoxy-4-methylbenzenemethanol ≥95%
α-Cyclopropyl-3-methoxy-4-methylbenzenemethanol

Precaución: For Research Use Only

Formula: 192.25 g/mol
MW: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD29965043
CAS Number: 1601947-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR