α-Cyclopropyl-2-methoxy-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259349-5G 259349-1G 259349-500MG
77628-258EA 1484.23 USD
77628-258 77641-542 77645-504
α-Cyclopropyl-2-methoxy-3-methylbenzenemethanol ≥95%
α-Cyclopropyl-2-methoxy-3-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34875792
CAS Number: 2352694-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR