α-Cyclopropyl-2,3-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259332-5G 259332-25G 259332-1G 259332-500MG
77632-310EA 1381 USD
77632-310 77654-502 77641-540 77645-502
α-Cyclopropyl-2,3-dimethylbenzenemethanol ≥95%
α-Cyclopropyl-2,3-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: 176.26 g/mol
MW: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD16301702
CAS Number: 1225604-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR