Benzenemethanol, α-cyclopropyl-2-ethyl- ≥95%

Supplier: AOBChem USA

Warning

259330-5G 259330-1G 259330-25G 259330-500MG
77632-308EA 1381 USD
77632-308 77637-520 77658-260 77649-582
Benzenemethanol, α-cyclopropyl-2-ethyl- ≥95%
Benzenemethanol, α-cyclopropyl-2-ethyl-

Precaución: For Research Use Only

Formula: 176.26 g/mol
MW: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD21328347
CAS Number: 1479255-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR