Benzenemethanol, α-cyclopropyl-2-(1-methylethyl)- ≥95%

Supplier: AOBChem USA

Warning

259324-5G 259324-1G 259324-500MG
77628-256EA 1484.23 USD
77628-256 77637-518 77649-580
Benzenemethanol, α-cyclopropyl-2-(1-methylethyl)- ≥95%
Benzenemethanol, α-cyclopropyl-2-(1-methylethyl)-

Precaución: For Research Use Only

Formula: 190.28 g/mol
MW: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD34875788
CAS Number: 1862776-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR