α-Cyclopropyl-3,4-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259316-5G 259316-1G 259316-25G 259316-500MG
77632-306EA 1206 USD
77632-306 77637-516 77658-258 77649-578
α-Cyclopropyl-3,4-dimethylbenzenemethanol ≥95%
α-Cyclopropyl-3,4-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11934736
CAS Number: 83949-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR