α-Cyclopropyl-3,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259302-5G 259302-25G 259302-1G 259302-500MG
77632-302EA 1206 USD
77632-302 77654-498 77641-536 77649-576
α-Cyclopropyl-3,5-dimethylbenzenemethanol ≥95%
α-Cyclopropyl-3,5-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD12767796
CAS Number: 1108697-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR