2,5-Dichloro-1-methylpyridin-4(1H)-one ≥97%

Supplier: AOBChem USA

Warning

25930-5G 25930-25G 25930-1G 25930-500MG
77632-304EA 919 USD
77632-304 77654-500 77641-534 77645-498
2,5-Dichloro-1-methylpyridin-4(1H)-one ≥97%
2,5-Dichloro-1-methylpyridin-4(1H)-one

Precaución: For Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24848705
CAS Number: 1449008-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR