α-Cyclopropyl-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259290-5G 259290-25G 259290-1G 259290-500MG
77632-300EA 1206 USD
77632-300 77654-496 77641-532 77649-574
α-Cyclopropyl-2-methoxybenzenemethanol ≥95%
α-Cyclopropyl-2-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11934799
CAS Number: 400613-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR